Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDA40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_119950.2 | NP_195502.2 | 1 | 2067 | 64 | |
NM_179183.2 | NP_849514.1 | 2 | 1994 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Both primers and probe map within a single exon |