Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160827.2 | NP_001154299.1 | 1 - 2 | 239 | 90 | |
NM_100011.4 | NP_171636.1 | 1 - 2 | 239 | 90 | ||
GenBank mRNA | BT025641.1 | - | 1 - 2 | 142 | 90 | |
Z48046.1 | - | 155 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |