Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331381.1 | NP_001322129.1 | 14 - 15 | 6448 | 84 | |
NM_001331382.1 | NP_001322128.1 | 14 - 15 | 6461 | 84 | ||
NM_001331383.1 | NP_001322130.1 | 14 - 15 | 6068 | 84 | ||
NM_100188.1 | NP_171805.1 | 14 - 15 | 5802 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |