Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ATG18G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100220.8 | NP_171837.7 | 7 - 8 | 2829 | 148 | |
GenBank mRNA | BT002410.1 | - | 7 - 8 | 2770 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |