Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100337.3 | NP_563711.1 | 2 - 3 | 2123 | 131 | |
GenBank mRNA | AB037271.1 | - | 2 - 3 | 1740 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |