Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100339.2 | NP_171954.1 | 14 - 15 | 1892 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |