Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001197984.1 | NP_001184913.1 | 41 - 42 | 5449 | 136 | |
NM_001331555.1 | NP_001322482.1 | 40 - 41 | 5663 | 136 | ||
NM_001331556.1 | NP_001322483.1 | 39 - 40 | 5497 | 136 | ||
NM_100436.5 | NP_563743.2 | 40 - 41 | 5746 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |