Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAUT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337197.1 | NP_001325285.1 | 2 - 3 | 198 | 97 | |
NM_001337198.1 | NP_001325284.1 | 2 - 3 | 150 | 97 | ||
NM_001337199.1 | NP_001325283.1 | 1 - 2 | 115 | 97 | ||
NM_130212.3 | NP_182171.1 | 1 - 2 | 115 | 97 | ||
GenBank mRNA | KJ676986.1 | - | 1 - 2 | 115 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |