Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DEG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001125072.1 | NP_001118544.1 | 5 - 6 | 582 | 75 | |
NM_130361.5 | NP_566115.1 | 5 - 6 | 635 | 75 | ||
GenBank mRNA | AF245171.1 | - | 5 - 6 | 566 | 75 | |
AF326865.1 | - | 5 - 6 | 508 | 75 | ||
AF349516.1 | - | 5 - 6 | 498 | 75 | ||
AY075700.1 | - | 5 - 6 | 508 | 75 | ||
AY102139.1 | - | 5 - 6 | 498 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |