Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100769.4 | NP_563831.1 | 1 - 2 | 208 | 66 | |
GenBank mRNA | AF202077.1 | - | 1 - 2 | 128 | 66 | |
AY037198.1 | - | 1 - 2 | 134 | 66 | ||
BT004554.1 | - | 1 - 2 | 104 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |