Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT1G10150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100889.4 | NP_563862.1 | 1 | 1681 | 156 | |
GenBank mRNA | AK226314.1 | - | 1 | 1515 | 156 |
Gene Symbol: PP2-A10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_148455.2 | NP_683296.1 | 3 | 713 | 156 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Both primers and probe map within a single exon |