Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NAT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001331912.1 | NP_001320848.1 | 4 - 5 | 693 | 68 | |
NM_100929.5 | NP_172524.1 | 4 - 5 | 776 | 68 | ||
GenBank mRNA | AY063935.1 | - | 4 - 5 | 523 | 68 | |
AY096464.1 | - | 4 - 5 | 504 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |