Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RECQ4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_100968.3 | NP_172562.2 | 3 - 4 | 484 | 68 | |
GenBank mRNA | AY120761.1 | - | 3 - 4 | 382 | 68 | |
BT010133.1 | - | 2 - 3 | 178 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |