Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scpl31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198028.1 | NP_001184957.1 | 8 - 9 | 1277 | 72 | |
NM_100981.4 | NP_172575.2 | 8 - 9 | 1482 | 72 | ||
GenBank mRNA | BT030381.1 | - | 8 - 9 | 1228 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |