Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUM12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345201.1 | NP_001318812.1 | 4 - 5 | 1816 | 85 | |
NM_001345202.1 | NP_001332601.1 | 4 - 5 | 1816 | 85 | ||
NM_001345203.1 | NP_001332600.1 | 4 - 5 | 1548 | 85 | ||
NM_001345204.1 | NP_001332599.1 | 4 - 5 | 1548 | 85 | ||
NM_001345205.1 | NP_001332598.1 | 3 - 4 | 1442 | 85 | ||
NM_125034.2 | NP_200462.2 | 3 - 4 | 1421 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |