Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ORP4C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037011.2 | NP_001032088.1 | 5 - 6 | 956 | 115 | |
NM_001345250.1 | NP_001329986.1 | 4 - 5 | 1363 | 115 | ||
NM_001345251.1 | NP_001329984.1 | 5 - 6 | 1154 | 115 | ||
NM_001345252.1 | NP_001329985.1 | 5 - 6 | 1154 | 115 | ||
NM_125106.3 | NP_200534.1 | 4 - 5 | 1363 | 115 | ||
GenBank mRNA | AK117264.1 | - | 4 - 5 | 914 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |