Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XTH25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125136.4 | NP_568859.2 | 1 - 2 | 338 | 79 | |
GenBank mRNA | AY125495.1 | - | 1 - 2 | 319 | 79 | |
AY143939.1 | - | 1 - 2 | 282 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |