Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125447.2 | NP_568922.1 | 1 - 2 | 339 | 82 | |
GenBank mRNA | AK117313.1 | - | 1 - 2 | 339 | 82 | |
AY087902.1 | - | 1 - 2 | 247 | 82 | ||
BT005266.1 | - | 1 - 2 | 184 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |