Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCA12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345489.1 | NP_001318859.1 | 4 - 5 | 732 | 148 | |
NM_001345490.1 | NP_001332309.1 | 4 - 5 | 817 | 148 | ||
NM_001345491.1 | NP_001332310.1 | 4 - 5 | 792 | 148 | ||
NM_001345492.1 | NP_001332311.1 | 4 - 5 | 792 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |