Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125591.3 | NP_201004.2 | 2 - 3 | 541 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |