Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_103814.4 | NP_175350.1 | 2 - 3 | 339 | 120 | |
GenBank mRNA | AF370302.1 | - | 2 - 3 | 227 | 120 | |
AY063089.1 | - | 1 - 2 | 60 | 120 | ||
AY124004.1 | - | 2 - 3 | 229 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Endogenous Controls Detect Genomic DNA Recommended Assay Arabidopsis thaliana Inventoried Probe spans exons |