Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128044.3 | NP_180058.1 | 5 - 6 | 1131 | 107 | |
GenBank mRNA | AK226395.1 | - | 5 - 6 | 1131 | 107 | |
BT002475.1 | - | 5 - 6 | 1131 | 107 | ||
BT006593.1 | - | 5 - 6 | 1067 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |