Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPHA-VPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128154.5 | NP_180165.1 | 6 - 7 | 1210 | 58 | |
GenBank mRNA | AY058055.1 | - | 6 - 7 | 1054 | 58 | |
AY090296.1 | - | 6 - 7 | 1046 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |