Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ER | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128190.3 | NP_180201.1 | 15 - 16 | 1921 | 136 | |
GenBank mRNA | AY035110.1 | - | 15 - 16 | 1230 | 136 | |
FJ708701.1 | - | 15 - 16 | 1154 | 136 | ||
U47029.1 | - | 15 - 16 | 1204 | 136 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |