Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDPMEK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128250.4 | NP_180261.1 | 4 - 5 | 473 | 121 | |
GenBank mRNA | AB037877.1 | - | 4 - 5 | 417 | 121 | |
AX765854.1 | - | 4 - 5 | 412 | 121 | ||
AY136394.1 | - | 4 - 5 | 429 | 121 | ||
BT000208.1 | - | 4 - 5 | 412 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |