Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001336102.1 | NP_001325161.1 | 15 - 16 | 1824 | 76 | |
NM_001336103.1 | NP_001325160.1 | 15 - 16 | 1824 | 76 | ||
NM_128270.3 | NP_180280.2 | 15 - 16 | 1824 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |