Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IPT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128335.2 | NP_565658.1 | 3 - 4 | 456 | 85 | |
GenBank mRNA | AB062609.1 | - | 3 - 4 | 368 | 85 | |
AF109376.2 | - | 3 - 4 | 430 | 85 | ||
BT001906.1 | - | 3 - 4 | 368 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |