Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AT2G28390 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128399.4 | NP_029426.1 | 10 - 11 | 1584 | 101 | |
GenBank mRNA | AK118037.1 | - | 10 - 11 | 1371 | 101 | |
AK175575.1 | - | 10 - 11 | 1371 | 101 | ||
AK175699.1 | - | 10 - 11 | 1371 | 101 | ||
AK176064.1 | - | 10 - 11 | 1381 | 101 | ||
AK176154.1 | - | 10 - 11 | 1381 | 101 | ||
AK221211.1 | - | 10 - 11 | 1371 | 101 | ||
AY056801.1 | - | 10 - 11 | 1364 | 101 | ||
AY133524.1 | - | 10 - 11 | 1294 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |