Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JOSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_128518.2 | NP_180525.1 | 3 - 4 | 648 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |