Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUR7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035662.2 | NP_001030739.1 | 8 - 9 | 1144 | 78 | |
NM_001338505.1 | NP_001319605.1 | 7 - 8 | 1317 | 78 | ||
NM_113005.4 | NP_188748.1 | 8 - 9 | 1220 | 78 | ||
GenBank mRNA | BT002734.1 | - | 7 - 8 | 928 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |