Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001203018.1 | NP_001189947.1 | 15 - 16 | 1488 | 71 | |
NM_113117.4 | NP_566700.1 | 15 - 16 | 1285 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |