Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SBP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113284.3 | NP_189022.1 | 1 - 2 | 376 | 99 | |
GenBank mRNA | AK117618.1 | - | 1 - 2 | 168 | 99 | |
BT005102.1 | - | 1 - 2 | 121 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |