Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PERK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113366.3 | NP_189098.1 | 6 - 7 | 1802 | 91 | |
GenBank mRNA | AY056788.1 | - | 6 - 7 | 1522 | 91 | |
AY059901.1 | - | 6 - 7 | 1517 | 91 | ||
AY093065.1 | - | 6 - 7 | 1515 | 91 | ||
AY128792.1 | - | 6 - 7 | 1486 | 91 | ||
BT008400.1 | - | 6 - 7 | 1486 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |