Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338695.1 | NP_001319633.1 | 4 - 5 | 792 | 117 | |
NM_113370.3 | NP_189102.3 | 4 - 5 | 824 | 117 | ||
GenBank mRNA | BT015406.1 | - | 4 - 5 | 762 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |