Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMKL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_113377.4 | NP_189109.1 | 1 - 2 | 1447 | 115 | |
GenBank mRNA | AY090945.1 | - | 1 - 2 | 1426 | 115 | |
X72863.1 | - | 1440 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Recommended Assay Arabidopsis thaliana Probe spans exons |