Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001338780.1 | NP_001327634.1 | 5 - 6 | 689 | 70 | |
NM_113514.4 | NP_189238.2 | 5 - 6 | 689 | 70 | ||
GenBank mRNA | AY150429.1 | - | 5 - 6 | 419 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |