Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125709.5 | NP_201119.1 | 1 - 2 | 421 | 84 | |
GenBank mRNA | AY882861.1 | - | 1 - 2 | 303 | 84 | |
BT014894.1 | - | 1 - 2 | 367 | 84 | ||
BT015022.1 | - | 1 - 2 | 303 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |