Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RMI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125749.5 | NP_201159.2 | 3 - 4 | 952 | 88 | |
GenBank mRNA | AY735746.1 | - | 3 - 4 | 917 | 88 | |
AY954880.1 | - | 3 - 4 | 854 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |