Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHR24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345616.1 | NP_001332133.1 | 6 - 7 | 1715 | 114 | |
NM_125791.3 | NP_201200.2 | 6 - 7 | 1817 | 114 | ||
GenBank mRNA | AF462829.1 | - | 6 - 7 | 1651 | 114 | |
AY142010.1 | - | 6 - 7 | 1551 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |