Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OPT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125837.4 | NP_201246.1 | 2 - 3 | 1103 | 66 | |
GenBank mRNA | AK227021.1 | - | 2 - 3 | 654 | 66 | |
AY099843.1 | - | 2 - 3 | 650 | 66 | ||
BT000339.1 | - | 2 - 3 | 576 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Arabidopsis thaliana Probe spans exons |