Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_125857.3 | NP_201266.1 | 6 - 7 | 1065 | 73 | |
GenBank mRNA | AB493812.1 | - | 6 - 7 | 899 | 73 | |
AY099684.1 | - | 6 - 7 | 926 | 73 | ||
BT000277.1 | - | 6 - 7 | 899 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Arabidopsis thaliana Probe spans exons |