Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AIL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001345694.1 | NP_001318882.1 | 6 - 7 | 1035 | 107 | |
NM_001345695.1 | NP_001330267.1 | 4 - 5 | 826 | 107 | ||
NM_001345696.1 | NP_001330268.1 | 4 - 5 | 826 | 107 | ||
NM_125949.7 | NP_201354.5 | 5 - 6 | 892 | 107 | ||
GenBank mRNA | AY560887.1 | - | 4 - 5 | 718 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |