Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332064.1 | NP_001322783.1 | 3 - 4 | 984 | 138 | |
NM_101193.3 | NP_563923.1 | 3 - 4 | 842 | 138 | ||
NM_101194.2 | NP_563924.1 | 3 - 4 | 749 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |