Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001332527.1 | NP_001320240.1 | 5 - 6 | 804 | 73 | |
NM_102025.3 | NP_564150.1 | 5 - 6 | 616 | 73 | ||
GenBank mRNA | AK228033.1 | - | 5 - 6 | 536 | 73 | |
BT006427.1 | - | 4 - 5 | 486 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |