Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPK29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334707.1 | NP_001319387.1 | 4 - 5 | 1674 | 116 | |
NM_001334708.1 | NP_001320822.1 | 4 - 5 | 1356 | 116 | ||
NM_106253.4 | NP_177731.2 | 3 - 4 | 640 | 116 | ||
NM_202421.3 | NP_974150.2 | 4 - 5 | 1356 | 116 | ||
GenBank mRNA | AY093022.1 | - | 3 - 4 | 570 | 116 | |
BT000401.1 | - | 3 - 4 | 492 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |