Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334912.1 | NP_001320544.1 | 18 - 19 | 2073 | 89 | |
NM_106618.5 | NP_565218.1 | 17 - 18 | 2245 | 89 | ||
GenBank mRNA | AY120774.1 | - | 17 - 18 | 1961 | 89 | |
BT000142.1 | - | 17 - 18 | 1931 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |