Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ELF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_106622.5 | NP_178091.2 | 8 - 9 | 1456 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |