Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP5K9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035589.3 | NP_001030666.1 | 2 - 3 | 1737 | 86 | |
NM_001202923.1 | NP_001189852.1 | 2 - 3 | 1667 | 86 | ||
NM_001337850.1 | NP_001327647.1 | 2 - 3 | 1681 | 86 | ||
NM_111827.3 | NP_187603.1 | 1 - 2 | 2533 | 86 | ||
GenBank mRNA | AJ810853.1 | - | 1 - 2 | 1267 | 86 | |
AK229815.1 | - | 2 - 3 | 1516 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |