Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDX26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_111898.4 | NP_187673.1 | 1 - 2 | 333 | 68 | |
GenBank mRNA | AF370187.1 | - | 1 - 2 | 233 | 68 | |
AY059145.1 | - | 1 - 2 | 233 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |