Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYCB1;3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001337935.1 | NP_001326714.1 | 2 - 3 | 545 | 141 | |
NM_111985.5 | NP_187759.2 | 2 - 3 | 545 | 141 | ||
GenBank mRNA | AK227951.1 | - | 2 - 3 | 502 | 141 | |
BT006437.1 | - | 2 - 3 | 411 | 141 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Arabidopsis thaliana Probe spans exons |